4-Chlorotetrahydropyran 96%

Leverandør: Thermo Fisher Scientific
Warning

434270010 434270050
ACRO434270010EA 830 NOK
ACRO434270010 ACRO434270050
4-Chlorotetrahydropyran 96%
4-Chlorotetrahydropyran
Formel: C₅H₉ClO
Kokepunkt: 150 °C (760 mmHg)
Tetthet: 1,12 g/cm³
Flash punkt: 46 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00074911
CAS-nummer: 1768-64-5
FN: 1993
ADR: 3,III

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to dark yellow Liquid
Infrared spectrum Conforms
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR