2-Amino-3-hydroxybenzoic acid 97%

Leverandør: Thermo Scientific
Warning

Synonymer: 3-Hydroxyanthranilic acid

434870010
ACRO434870010EA 2650 NOK
ACRO434870010
2-Amino-3-hydroxybenzoic acid 97%
2-Amino-3-hydroxybenzoic acid
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Kokepunkt: 357 °C (1013 hPa)
Smeltepunkt: 224…231 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007700
CAS-nummer: 548-93-6

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to orange Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR