BippyPhos 97%

Leverandør: Thermo Scientific

Synonymer: 5-(DI-T-BUTYLPHOSPHINO)-1',3',5'-TRIPHENYL-1,4'-BI-1H-PYRAZOLE, 5-(Di-tert-butylphosphino)-1', 3', 5'-triphenyl-1'H-[1,4']bipyrazole, 5-(di-tert-Butylphosphino)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole, 5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole

435160010
ACRO435160010EA 3525 NOK
ACRO435160010
BippyPhos 97%
BippyPhos
Formel: C₃₂H₃₅N₄P
MW: 506,634 g/mol
Smeltepunkt: 169…173 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09038440
CAS-nummer: 894086-00-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to tan Crystalline powder or solid
Infrared spectrum Conforms
NMR ≥96.0 %
Specific optical rotation +64° to +68° (25°C, 589 nm) (c=5.5, CH2Cl2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR