Tetrabenzyl pyrophosphate 98%

Leverandør: Thermo Fisher Scientific
Danger

436750010
ACRO436750010EA 2200 NOK
ACRO436750010
Tetrabenzyl pyrophosphate 98%
Tetrabenzyl pyrophosphate
Formel: C₂₈H₂₈O₇P₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00051941
CAS-nummer: 990-91-0
FN: 3261
ADR: 8,II

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 59 to 65 °C
Water ≤0.1 %
HPLC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR