Norfloxacin, 98%, analytical standard

Leverandør: Thermo Scientific

445260050
ACRO445260050EA 1575 NOK
ACRO445260050
Norfloxacin, 98%, analytical standard
Standarder Organiske standarder

Bestill nå

SPESIFIKASJONER

Beskrivelse Norfloxacin
CAS-nr 70458-96-7
Formel C₁₆H₁₈FN₃O₃
Smeltepunkt 218…226 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR