Natriumklorid 99.5% for HPLC

Leverandør: Thermo Fisher Scientific

446212500
ACRO446212500EA 1625 NOK
ACRO446212500
Natriumklorid 99.5% for HPLC
Natriumklorid
Formel: NaCl
Kokepunkt: 1461 °C (760 mmHg)
Smeltepunkt: 801 °C
Tetthet: 2,16 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003477
CAS-nummer: 7647-14-5

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Titration Argentometric ≥99.4 %
UV at 500 nm A: ≤0.01
pH 5.0 to 9.0

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR