Natriumacetat, vannfri 99% for HPLC

Leverandør: Thermo Fisher Scientific

446880500 446882500
ACRO446880500EA 830 NOK
ACRO446880500 ACRO446882500
Natriumacetat, vannfri 99% for HPLC
Natriumacetat
Formel: H₃CCOONa
Kokepunkt: 881,4 °C (1013 hPa)
Smeltepunkt: 324 °C
Tetthet: 1,528 g/cm³ (20 °C)
Flash punkt: >250 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012459
CAS-nummer: 127-09-3

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Loss on drying ≤1 %
UV (0.5 M in water)
Titration with HClO4 ≥98.5 %
Insoluble matter ≤0.005 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR