Rhodamine WT 20% in water

Leverandør: Thermo Fisher Scientific

446970010 446971000
ACRO446970010EA 4775 NOK
ACRO446970010 ACRO446971000
Rhodamine WT 20% in water
Rhodamine WT
Formel: C₂₉H₂₉ClN₂Na₂O₅
Tetthet: 1,16 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD01728973
CAS-nummer: 37299-86-8

Bestill nå

Spesifikasjon Testresultater

Appearance Dark red Liquid
UV Lambda max 553 to 557 nm
Strength 19.5 to 21 % (dye content)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR