Karbondisulfid 99.9% ACS

Leverandør: Thermo Fisher Scientific
Danger

447035000
ACRO447035000EA 965 NOK
ACRO447035000
Karbondisulfid 99.9% ACS
Karbondisulfid
Formel: CS₂
Kokepunkt: 46 °C (760 mmHg)
Smeltepunkt: –111 °C
Tetthet: 1,262 g/cm³
Flash punkt: –30 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011321
CAS-nummer: 75-15-0
FN: 1131
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Water ≤0.05 %
GC ≥99.9 %
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Sulfur dioxide (SO2) ≤2.5 ppm
Hydrogen sulfide (H2S) ≤1.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR