Kaliumhydrogenortofosfat, vannfri 99% for HPLC

Leverandør: Thermo Fisher Scientific

447362500
ACRO447362500EA 3175 NOK
ACRO447362500
Kaliumhydrogenortofosfat, vannfri 99% for HPLC
Kaliumhydrogenortofosfat
Formel: K₂HPO₄
Smeltepunkt: 340 °C
Tetthet: 2,3 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011383
CAS-nummer: 7758-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder or crystals or granules
Loss on drying ≤1 %
Solubility Clear colorless
UV at 500 nm A: ≤0.02
pH 8.5 to 9.6 (10 % in water)
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR