Magnesiumnitrat heksahydrat, extra pure

Leverandør: Thermo Fisher Scientific

447540010 447540050 447540250
ACRO447540010EA 770 NOK
ACRO447540010 ACRO447540050 ACRO447540250
Magnesiumnitrat heksahydrat, extra pure
Magnesiumnitrat heksahydrat
Formel: Mg(NO₃)₂·6H₂O
Kokepunkt: 330 °C (760 mmHg)
Smeltepunkt: 89 °C
Tetthet: 1,63 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149779
CAS-nummer: 13446-18-9
ADR: 5.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystals
Titration Complexometric 98 to 102 %
pH 5 to 8.2 (5 % in water)
Chloride (Cl) ≤0.001 %
Sulfate (SO4) ≤0.005 %
Iron (Fe) ≤0.001 %
Heavy metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR