Perfluorokerosene 50% PFK low boiling, boiling_high Mass spectrometry

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: PFK

450130010
ACRO450130010EA 5625 NOK
ACRO450130010
Perfluorokerosene 50% PFK low boiling, boiling_high Mass spectrometry
Perfluorokerosene
Kokepunkt: 180 °C
Tetthet: 1.89 g/cm³
Flash punkt: 46 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00132835
FN: 1993
ADR: 3 III

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Suitability (for mass analysis) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR