Choline hydrogen tartrate 97%

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate

450221000 450225000
ACRO450221000EA 805 NOK
ACRO450221000 ACRO450225000
Choline hydrogen tartrate 97%
Choline hydrogen tartrate
Formel: C₉H₁₉NO₇
Smeltepunkt: 151…153 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00036332
CAS-nummer: 87-67-2

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR