Perchloric acid 60% in water for analysis

Leverandør: Thermo Fisher Scientific
Danger

452850010
ACRO452850010EA 2450 NOK
ACRO452850010
Perchloric acid 60% in water for analysis
Perchloric acid
Formel: HClO₄
Kokepunkt: 203 °C (760 mmHg)
Smeltepunkt: –18 °C
Tetthet: 1,66 g/cm³
Flash punkt: 113 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011325
CAS-nummer: 7601-90-3
FN: 1873
ADR: 5.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Clear colorless Liquid
Color scale ≤10 APHA
Titration with NaOH 60.0 to 65.0 %
Chloride (Cl) ≤3 ppm
Chlorate (ClO3) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Silicate (as SiO2) ≤5 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Copper (Cu) ≤0.1 ppm
Iron (Fe) ≤1 ppm
Lead (Pb) ≤0.05 ppm
Magnesium (Mg) ≤0.5 ppm
Nickel (Ni) ≤0.1 ppm
Nitrogen (N) ≤10 ppm
Potassium (K) ≤0.1 ppm
Sodium (Na) ≤0.5 ppm
Zinc (Zn) ≤0.1 ppm

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR