Cefazolin sodium salt 98%

Leverandør: Thermo Fisher Scientific
Danger

455210010 455210050
ACRO455210010EA 1350 NOK
ACRO455210010 ACRO455210050
Cefazolin sodium salt 98%
Cefazolin sodium salt
Formel: C₁₄H₁₄N₈NaO₄S₃
Smeltepunkt: 170…177 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00056883
CAS-nummer: 27164-46-1

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 170 to 177 °C
Water ≤6 %
HPLC ≥97.5 %
Specific optical rotation -24° to -10° (20 °C, 589 nm) (c=1,H2O)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR