Penicillin V potassium salt 99%

Leverandør: Thermo Fisher Scientific
Danger

455230250 455230050
ACRO455230250EA 1675 NOK
ACRO455230250 ACRO455230050
Penicillin V potassium salt 99%
Penicillin V potassium salt
Formel: C₁₆H₁₇KN₂O₅S
Smeltepunkt: 197…202 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00051771
CAS-nummer: 132-98-9

Bestill nå

Spesifikasjon Testresultater

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Specific optical rotation +220° to +235° (20°C, 589 nm) (c=1, H2O)

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR