Gentamicin sulphate

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

455310010 455310050 455310250
ACRO455310010EA 560 NOK
ACRO455310010 ACRO455310050 ACRO455310250
Gentamicin sulphate
Gentamicin sulphate
Smeltepunkt: 218…237 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00270181
CAS-nummer: 1405-41-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Powder
Infrared spectrum Conforms
Assay ≥590 µg/mg (On dried basis)
Loss on drying ≤18.0 % (Vacuum, 110°C, 3h)
Sulfated ash ≤1.0 %
pH 3.5 to 5.5 (c=4, water)
Specific optical rotation (On dried basis)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR