Imipenem monohydrate 98%

Leverandør: Thermo Fisher Scientific

455320010 455322500
ACRO455320010EA 9775 NOK
ACRO455320010 ACRO455322500
Imipenem monohydrate 98%
Imipenem monohydrate
Formel: C₁₂H₁₇N₃O₄S
Smeltepunkt: 193…198 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD09753321
CAS-nummer: 74431-23-5

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to light yellow Powder
Infrared spectrum Conforms
Water 5 to 8 %
HPLC ≥97.5 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR