Streptomycin sulphate

Leverandør: Thermo Fisher Scientific
Warning

455340050 455340250 455341000
ACRO455340050EA 475 NOK
ACRO455340050 ACRO455340250 ACRO455341000
Streptomycin sulphate
Streptomycin sulphate
Formel: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00037023
CAS-nummer: 3810-74-0

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Loss on drying ≤7 %
Sulfate (SO4) 18.0 to 21.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR