Carbenicillin disodium salt 90%

Leverandør: Thermo Fisher Scientific
Danger

455360010 455360050
ACRO455360010EA 1950 NOK
ACRO455360010 ACRO455360050
Carbenicillin disodium salt 90%
Carbenicillin disodium salt
Formel: C₁₇H₁₆N₂O₆S·2Na
Kokepunkt: 738 °C
Smeltepunkt: >200 °C
Flash punkt: 400 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00077683
CAS-nummer: 4800-94-6

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤6 %
Solubility (5 % in water) Clear to light yellow
HPLC ≥88 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR