Lutein 90%

Leverandør: Thermo Fisher Scientific

Synonymer: E 161b, (6'R)-β,ε-carotene-3R,3'R-diol, Xanthophyll, (1R,4R)-4-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-18-((R)-4-Hydroxy-2,6,6-trimethylcyclohex-1-en-1-yl)-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaen-1-yl)-3,5,5-trimethylcyclohex-2-enol

456160010 456160050
ACRO456160010EA 4225 NOK
ACRO456160010 ACRO456160050
Lutein 90%
Lutein
Formel: C₄₀H₅₆O₂
MW: 568.88 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD08435941
CAS-nummer: 127-40-2

Bestill nå

Spesifikasjon Testresultater

Appearance Yellow to red Powder
Infrared spectrum Conforms
HPLC ≥88 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR