Salbutamol sulphate 98%

Leverandør: Thermo Fisher Scientific
Warning

456460010 456460050
ACRO456460010EA 1325 NOK
ACRO456460010 ACRO456460050
Salbutamol sulphate 98%
Salbutamol sulphate
Formel: (C₁₃H₂₁NO₃)₂·H₂SO₄
Smeltepunkt: 180 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00055200
CAS-nummer: 51022-70-9

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR