2'-Deoxycytidine hydrochloride 98%

Leverandør: Thermo Fisher Scientific

458510010 458510050 458510250
ACRO458510010EA 690 NOK
ACRO458510010 ACRO458510050 ACRO458510250
2'-Deoxycytidine hydrochloride 98%
2'-Deoxycytidine hydrochloride
Formel: C₉H₁₄ClN₃O₄
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00012840
CAS-nummer: 3992-42-5

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤8 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR