7-Ethyl-10-hydroxycamptothecin 98%

Leverandør: Thermo Fisher Scientific
Danger

459090010 459091000
ACRO459090010EA 4700 NOK
ACRO459090010 ACRO459091000
7-Ethyl-10-hydroxycamptothecin 98%
7-Ethyl-10-hydroxycamptothecin
Formel: C₂₂H₂₀N₂O₅
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00871873
CAS-nummer: 86639-52-3
FN: 1544
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Appearance Off-white to yellow Powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR