Benzbromarone 98%

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: (3,5-Dibromo-4-hydroxyphenyl)(2-ethylbenzofuran-3-yl)methanone, 3-(3,5-Dibromo-4-hydroxybenzoyl)-2-ethylbenzofuran

460900010 460900050
ACRO460900010EA 520 NOK
ACRO460900010 ACRO460900050
Benzbromarone 98%
Benzbromarone
Formel: C₁₇H₁₂Br₂O₃
MW: 424.09 g/mol
Smeltepunkt: 152 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00078962
CAS-nummer: 3562-84-3
FN: 2811
ADR: 6.1 III

Bestill nå

Spesifikasjon Testresultater

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
Loss on drying ≤0.5 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR