Quetiapine hemifumarate

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: ICI-204636, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol bis[(2E)-2-butenedioate], Seroquel

462400050 462400250
ACRO462400050EA 980 NOK
ACRO462400050 ACRO462400250
Quetiapine hemifumarate
Quetiapine hemifumarate

  • Bioactive

Formel: C₂₁H₂₅N₃O₂S
MW: 441.55 g/mol
Smeltepunkt: 172…176 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD03423782
CAS-nummer: 111974-72-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 172 to 176 °C
Loss on drying ≤0.5 % (105 °C)
Solubility (2 % in DMSO) Clear colorless to light yellow solution
Titration with HClO4 99 to 101 % (quetiapine hemifumarate dry substance)
Titration with NaOH 12.5 to 13.8 % (fumaric acid dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR