Nebivolol hydrochloride

Leverandør: Thermo Fisher Scientific
Warning

462492500
ACRO462492500EA 2150 NOK
ACRO462492500
Nebivolol hydrochloride
Nebivolol hydrochloride

  • Bioactive

Formel: C₂₂H₂₅F₂NO₄
Smeltepunkt: 226 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD07357273
CAS-nummer: 152520-56-4
FN: 3077
ADR: 9,III

Bestill nå

Spesifikasjon Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
HPLC ≥97.5 % (mixture of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR