Meropenem trihydrate

Leverandør: Thermo Fisher Scientific
Warning

462810010 462810050
ACRO462810010EA 1250 NOK
ACRO462810010 ACRO462810050
Meropenem trihydrate
Meropenem trihydrate

  • Bioactive

Formel: C₁₇H₃₁N₃O₈S
Kokepunkt: 627 °C
Flash punkt: >110 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD08600005
CAS-nummer: 119478-56-7

Bestill nå

Spesifikasjon Testresultater

Appearance Almost white to light yellow Powder or crystals
Infrared spectrum Conforms
Water ≤15 %
HPLC ≥94 %
Specific optical rotation -17° to -21° (20°C, 589 nm) (0.5 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR