Desloratadine

Leverandør: Thermo Scientific

Synonymer: 8-Chloro-11-(piperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine

462920010
ACRO462920010EA 2550 NOK
ACRO462920010
Desloratadine
Desloratadine

  • Bioactive

Formel: C₁₉H₁₉ClN₂
MW: 310,83 g/mol
Smeltepunkt: 150 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00871949
CAS-nummer: 100643-71-8

Bestill nå

Spesifikasjon Testresultater

Appearance White to almost white Powder
Infrared spectrum Conforms
Melting point 150 to 159 °C
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR