Tilmicosin phosphate

Leverandør: Thermo Fisher Scientific
Danger

463270010
ACRO463270010EA 4125 NOK
ACRO463270010
Tilmicosin phosphate
Tilmicosin phosphate

  • Bioactive

Formel: C₄₆H₈₀N₂O₁₃·H₃PO₄
MW: 967.13 g/mol
Smeltepunkt: 130…135 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD06797805
CAS-nummer: 137330-13-3

Bestill nå

Spesifikasjon Testresultater

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥70 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR