Triptolide

Leverandør: Thermo Fisher Scientific
Danger

466840250
ACRO466840250EA 4650 NOK
ACRO466840250
Triptolide
Triptolide
Formel: C₂₀H₂₄O₆
MW: 360.41 g/mol
Smeltepunkt: 226°C…227°C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00210565
CAS-nummer: 38748-32-2

Bestill nå

Spesifikasjon Testresultater

Appearance White to almost white powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥ 97.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR