Sodium Standard Solution, 10000 mg/L Na in dil. Nitric Acid Standard for ICP-MS
Leverandør: CPACHEM
M135.2NP.L1
ACSDM135.2NP.L1EA 3050 NOK
ACSDM135.2NP.L1
Sodium Standard Solution, 10000 mg/L Na in dil. Nitric Acid Standard for ICP-MS
Standarder Kationstandarder Sodium Standards
Na 10000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Sertifiseringer: ISO 17034, ISO 17025, ISO 9001.