Germanium Standard Solution, 1000 mg/L Ge in Diluted HNO₃ and HF for ICP-MS
Leverandør: CPACHEM
M222.2N2FP.L1
ACSDM222.2N2FP.L1EA 1975 NOK
ACSDM222.2N2FP.L1
Germanium Standard Solution, 1000 mg/L Ge in Diluted HNO₃ and HF for ICP-MS
Standarder Kationstandarder Germanium Standards