Terbium Standard Solution, 1000 mg/L Tb in dil. Nitric Acid Standard for ICP-MS
Leverandør: CPACHEM
M258.2NP.L1
ACSDM258.2NP.L1EA 2100 NOK
ACSDM258.2NP.L1
Terbium Standard Solution, 1000 mg/L Tb in dil. Nitric Acid Standard for ICP-MS
Standarder Kationstandarder Terbium Standards
Tb 1000 mg/L in diluted nitric acid for ICP/MS.
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.
Sertifiseringer: ISO 17034, ISO 17025, ISO 9001.