4-Nonylphenol, ethoxylated

Leverandør: CPACHEM
Danger

Synonymer: 4-Nonylphenyl-polyethylene glycol

SB37640.100MG
ACSDSB37640.100MGEA 1575 NOK
ACSDSB37640.100MG
4-Nonylphenol, ethoxylated
4-Nonylphenol, ethoxylated
Smeltepunkt: 57 °C
Tetthet: 1,6 g/cm³ at 20 °C
Flash punkt: >93 °C
MDL nummer: MFCD00132851
CAS-nummer: 9016-45-9
FN: 3082
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR