Chromium ICP and ICP-MS CRM, Chromium (III) 1000 mg/L in Diluted Nitric Acid
Leverandør: CPACHEM
SP073.2N.L1
ACSDSP073.2N.L1EA 1800 NOK
ACSDSP073.2N.L1
Chromium ICP and ICP-MS CRM, Chromium (III) 1000 mg/L in Diluted Nitric Acid
Standarder Kationstandarder Chromium Standards
Shelf life on ship date: 36 months - store under normal laboratory conditions, at temperatures between 15 to 25 °C.