Anti-PsbA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Agrisera
AS05084A-HRP
AGRIAS05084A-HRPEA 5650 NOK
AGRIAS05084A-HRP
Anti-PsbA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
The psbA gene has been cloned from many species of plants, green algae, and cyanobacteria. The psbA gene is located in the chloroplast genome and encodes for the D1 protein, a core component of Photosystem II. PsbA/D1 is rapidly cycled under illumination in all oxygenic photobionts. Tracking PsbA pools using the Global PsbA antibody can show the functional content of Photosystem II in a wide range of samples. Alternative names: 32 kDa thylakoid membrane protein, photosystem II protein D1.,The psbA gene has been cloned from many species of plants, green algae, and cyanobacteria. The psbA geneMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol PSBA
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering HRP (Horseradish Peroxidase)
Reaktivitet Zea mays,A. thaliana,H. vulgare
Vert Rabbit
Isotype
Western blot Yes
Immunogen KLH-conjugated synthetic peptide derived from available plant, algal and cyanobacterial PsbA sequences, including Arabidopsis thaliana
UniProtKB A4QJR4

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR