Anti-PsaH Rabbit Polyclonal Antibody

Leverandør: Agrisera
AS06105
AGRIAS06105EA 3850 NOK
AGRIAS06105
Anti-PsaH Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PSAH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, N. tabacum, Spinacia oleracea
Bestill nå

SPESIFIKASJONER

Antigensymbol PSAH
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Spinacia oleracea,A. thaliana,N. tabacum
Vert Rabbit
Isotype
Western blot Yes
Immunogen KLH-conjugated synthetic peptide derived from the protein sequence of Arabidopsis thaliana for PsaH1(At3g16240) and PsaH2 (At1g52230). This peptide sequence is quite conserved in some dicots but not in monocots.
Kyssabsorpsjon No
Lagringstemp –20 °C
Frakttemp. Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR