Anti-SUMO3 Rabbit Polyclonal Antibody

Leverandør: Agrisera
AS08349
AGRIAS08349EA 3850 NOK
AGRIAS08349
Anti-SUMO3 Rabbit Polyclonal Antibody
Antistoffer
SUMO3 (Small ubiquitin-related modifier 3) is a small polypeptide that becomes covalently attached to various intracellular proteins leading to their post-translational modification. ,SUMO3 (Small ubiquitin-related modifier 3) is a small polypeptide that becomes covalently attached to various intracellular proteins leading to their post-translational modification.

Type: Primary
Antigen: SUMO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR