Anti-IgY Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: Agrisera
AS101104
AGRIAS101104EA 1975 NOK
AGRIAS101104
Anti-IgY Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken
Bestill nå

SPESIFIKASJONER

Antigensymbol IgY
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering TRITC (Tetramethylrhodamine Isothiocyanate)
Reaktivitet Chicken
Vert Goat
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Immunogen Purified Chicken IgY, whole molecule
Rensing purified
Kyssabsorpsjon No
Lagringstemp +2°C to +8 °C
Frakttemp. Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR