Anti-IgG (H+L) Chicken Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: Agrisera
AS101137
AGRIAS101137EA 2975 NOK
AGRIAS101137
Anti-IgG (H+L) Chicken Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse, Rabbit, Sheep
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR