Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: Agrisera
AS101197
AGRIAS101197EA 2250 NOK
AGRIAS101197
Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG F(c)
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering TRITC (Tetramethylrhodamine Isothiocyanate)
Reaktivitet Human
Vert Goat
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Immunogen Purified Human IgG, Fc fragment
Rensing purified
Kyssabsorpsjon No
Lagringstemp +2°C to +8 °C
Frakttemp. Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR