Anti-IgG F(c) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Agrisera
AS101236
AGRIAS101236EA 2975 NOK
AGRIAS101236
Anti-IgG F(c) Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR