Anti-IgG F(c) Rabbit Polyclonal Antibody (Biotin)

Leverandør: Agrisera
AS101241
AGRIAS101241EA 2975 NOK
AGRIAS101241
Anti-IgG F(c) Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rabbit
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG F(c)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet Rabbit
Vert Rabbit
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Immunogen Purified Goat IgG, Fc fragment
Rensing purified
Kyssabsorpsjon No
Lagringstemp +2°C to +8 °C
Frakttemp. Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR