Anti-IgG (H+L) Donkey Polyclonal Antibody (Biotin)

Leverandør: Agrisera
AS101258
AGRIAS101258EA 2975 NOK
AGRIAS101258
Anti-IgG (H+L) Donkey Polyclonal Antibody (Biotin)
Antistoffer
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR