Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Agrisera
AS101282
AGRIAS101282EA 2975 NOK
AGRIAS101282
Anti-IgG (H+L) Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Donkey
Isotype:
Reactivity: Mouse
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Environmentally Preferable
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering FITC (Fluorescein Isothiocyanate)
Reaktivitet Mouse
Vert Donkey
Isotype
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Immunogen Purified Rat IgG, whole molecule
Rensing purified
Kyssabsorpsjon No
Lagringstemp +2°C to +8 °C
Frakttemp. Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR