Anti-IgM µ Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: Agrisera
AS101290
AGRIAS101290EA 1900 NOK
AGRIAS101290
Anti-IgM µ Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol IgM µ
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering TRITC (Tetramethylrhodamine Isothiocyanate)
Reaktivitet Human
Vert Goat
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Immunogen Purified Human IgM (µ chain)
Rensing purified
Kyssabsorpsjon No
Lagringstemp +2°C to +8 °C
Frakttemp. Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR