Anti-IgG (H+L) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Leverandør: Agrisera
AS101421
AGRIAS101421EA 2975 NOK
AGRIAS101421
Anti-IgG (H+L) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype:
Reactivity: Llama
Bestill nå

SPESIFIKASJONER

Antigensymbol IgG (H+L)
Antistofftype Secondary
Klonalitet Polyclonal
Konjugering TRITC (Tetramethylrhodamine Isothiocyanate)
Reaktivitet Llama
Vert Goat
ELISA Yes
ImmunoKjemi Yes
Flowcytometri Yes
Immunogen Purified llama IgG (H&L) AAQ19986
Rensing purified
Kyssabsorpsjon No
Lagringstemp +2°C to +8 °C
Frakttemp. Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR