Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Leverandør: Agrisera
AS101502
AGRIAS101502EA 2475 NOK
AGRIAS101502
Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antistoffer
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR