Anti-IgE epsilon Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: Agrisera
AS10750
AGRIAS10750EA 3150 NOK
AGRIAS10750
Anti-IgE epsilon Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Type: Secondary
Antigen: IgE epsilon
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR